Cynomolgus Rhesus macaque TIGIT, His Tag (HPLC-verified) (TIT-C5223) is expressed from human 293 cells (HEK293). It contains AA Met 89 - Ile 208 (Accession XP015300911.1). In the region Met 89 - Ile 208, the AA sequence of Cynomolgus and Rhesus ma e 1 * 50 µG
Lieferant: ACROBIOSYSTEMS INC