Recombinat Protein,Mouse Siglec-2 CD22 Protein, His Tag,Mouse Siglec-2, His Tag (SI2-M52Ha) is expressed from human 293 cells (HEK2 93). It contains AA Ser 22 - Arg 702 (Accession P35329-1).This protein carries a polyhistidine tag at the C-terminus. 1 * 100 µG
Lieferant: ACROBIOSYSTEMS INC