Recombinat Protein,Mouse ROR2 NTRKR2 Protein, Fc Tag,Mouse ROR2, Fc Tag (RO2-M5253) is expressed from human 293 cells (HEK293). It contains AA Glu 34 - Gly 403 (Accession Q9Z138-1).This protein carries a human IgG1 Fc tag at the C-terminus. The prot 1 * 100 µG
Lieferant: ACROBIOSYSTEMS INC