Recombinat Protein,Biotinylated Human NTB-A SLAMF6 Protein, His,Avitag™,Biotinylated Human NTB-A, His,Avitag (NTA-H82E6) is express ed from human 293 cells (HEK293). It contains AA Gln 22 - Met 226 (Accession Q96DU3-1).This protein carries a polyhis 1 * 25 µG
Lieferant: ACROBIOSYSTEMS INC