Drucken
FITC-Labeled Human Mesothelin (296-580), Fc Tag (Cat. No. MSN-HF253) is expressed from human HEK293 cells. It contains AA Glu 296 - Gly 580 (Accession AAH09272.1). It is the FITC labeled form of Human Mesothelin (296-580), Fc Tag (Cat. No. MSN-H5253 1 * 25 µG
Lieferant: ACROBIOSYSTEMS INC