Recombinat Protein,Human Lumican Protein, His Tag,Human Lumican, His Tag (LUM-H5227) is expressed from human 293 cells (HEK293). It contains AA Gln 19 - Asn 338 (Accession NP002336).This protein carries a polyhistidine tag at the C-terminus. The pro 1 * 100 µG
Lieferant: ACROBIOSYSTEMS INC