Biotinylated Human LAP (TGF-beta 1), His,Avitag (LAP-H82Q6) is expressed from human 293 cells (HEK293). It contains AA Leu 30 - Arg 278 (Accession P01137-1 (C33S)). 1 * 25 µG
Lieferant: ACROBIOSYSTEMS INC