Recombinat Protein,Human IL-18BP Protein, Fc Tag,Human IL-18BP, Fc Tag (ILP-H5253) is expressed from human 293 cells (HEK293). It co ntains AA Thr 31 - Gly 194 (Accession NP001138529).This protein carries a human IgG1 Fc tag at the C-terminus. The p 1 * 200 µG
Lieferant: ACROBIOSYSTEMS INC