Recombinat Protein,Human IL-1 Rrp2 IL-1 R6 Protein, His Tag,Human IL-1 Rrp2, His Tag (IL2-H52H6) is expressed from human 293 cells (HEK293). It contains AA Asp 20 - Arg 335 (Accession Q9HB29-1).This protein carries a polyhistidine tag at the C-termi 1 * 100 µG
Lieferant: ACROBIOSYSTEMS INC