Recombinat Protein,Mouse IL-10 Protein, His Tag,Mouse IL-10, His Tag (IL0-M4248) is expressed from human 293 cells (HEK293). It cont ains AA Ser 19 - Ser 178 (Accession NP034678).This protein carries a polyhistidine tag at the N-terminus. The protei 1 * 20 µG
Lieferant: ACROBIOSYSTEMS INC