Recombinat Protein,Human ICAM-1 CD54 Protein, His Tag,Human ICAM-1, His Tag (IC1-H52H5) is expressed from human 293 cells (HEK293). It contains AA Gln 28 - Glu 480 (Accession P05362-1).This protein carries a polyhistidine tag at the C-terminus. The 1 * 100 µG
Lieferant: ACROBIOSYSTEMS INC