Biotinylated Influenza A AGuinea fowlHong KongWF1099(H9N2) HA, Avitag,His Tag (HA1-V82E1) is expressed from human 293 cells (HEK293) . It contains AA Asp 19 - Lys 523 (Accession AAO46082.1). 1 * 25 µG
Lieferant: ACROBIOSYSTEMS INC