Recombinat Protein,Influenza A AThailand1(KAN-1)2004 (H5N1) Neuraminidase (NA) Protein, His Tag,Influenza A AThailand1(KAN-1)2004 (H 5N1) NA, His Tag (H54-V5247) is expressed from human 293 cells (HEK293). It contains AA His 36 - Lys 449 (Accession o 1 * 100 µG
Lieferant: ACROBIOSYSTEMS INC