Recombinat Protein,Biotinylated Human GITR TNFRSF18 Protein, Fc,Avitag™,Biotinylated Human GITR, Fc,Avitag (GIR-H82F7) is expressed from human 293 cells (HEK293). It contains AA Gln 26 - Glu 161 (Accession Q9Y5U5-1).This protein carries a human IgG e 1 * 25 µG
Lieferant: ACROBIOSYSTEMS INC