MABSol® Biotinylated Cynomolgus Rhesus macaque FcRn Heterodimer Protein, His,AvitagStrep II Tag (BLI verified) (FCM-C82W5) is expre ssed from human HEK293 cells. It contains AA Ala 24 - Ser 297 (FCGRT) Ile 21 - Met 119 (B2M) (Accession Q8SPV9-1 (FC T 1 * 25 µG
Lieferant: ACROBIOSYSTEMS INC