Biotinylated Cynomolgus Rhesus macaque FcRn Heterodimer Protein, His,Avitag (SPR verified), is expressed from human 293 cells (HEK2 93). It contains AA Ala 24 - Ser 297 (FCGRT) Ile 21 - Met 119 (B2M) (Accession Q8SPV9-1 (FCGRT) Q8SPW0-1 (B2M)). In a 1 * 25 µG
Lieferant: ACROBIOSYSTEMS INC