Recombinat Protein,Human FAM171B Protein, His Tag,Human FAM171B, His Tag (FAB-H5220) is expressed from human 293 cells (HEK293). It contains AA Glu 33 - Thr 355 (Accession AAH60872).This protein carries a polyhistidine tag at the C-terminus. The pro 1 * 100 µG
Lieferant: ACROBIOSYSTEMS INC