Recombinat Protein,Human LSECtin CLEC4G Protein, Fc Tag,Human LSECtin, Fc Tag (CLG-H5250) is expressed from human 293 cells (HEK293 ). It contains AA Lys 55 - Cys 293 (Accession Q6UXB4).This protein carries a human IgG1 Fc tag at the N-terminus. The 1 * 50 µG
Lieferant: ACROBIOSYSTEMS INC