Recombinat Protein,Human LILRB4 CD85k ILT3 Protein, Mouse IgG2a Fc Tag, low endotoxin,Human LILRB4, Mouse IgG2a Fc Tag, low endoto xin (CDK-H5259) is expressed from human 293 cells (HEK293). It contains AA Gln 22 - Glu 259 (Accession AAH26309.1).Thi 1 * 100 µG
Lieferant: ACROBIOSYSTEMS INC