Biotinylated Mouse CD32b, Avitag,His Tag (SPR BLI verified) (CDB-M82E8) is expressed from human 293 cells (HEK293). It contains AA Thr 40 - Arg 217 (Accession NP001070657.1). 1 * 25 µG
Lieferant: ACROBIOSYSTEMS INC