Recombinat Protein,Mouse Glycophorin A CD235a Protein, Fc,His Tag,Mouse Glycophorin A, Fc,His Tag (CDA-M526x) is expressed from hum an 293 cells (HEK293). It contains AA Met 1 - Val 108 (Accession P14220).This protein carries a human IgG1 Fc tag at u 1 * 50 µG
Lieferant: ACROBIOSYSTEMS INC