Biotinylated Human CD16a (F176), Avitag,His Tag (SPR  BLI verified) (CDA-H82E8) is expressed from human 293 cells (HEK293). It conta ins AA Gly 17 - Gln 208 (Accession P08637-1). 1 * 25 µG
Lieferant: ACROBIOSYSTEMS INC