Biotinylated Cynomolgus CD32a, His,Avitag (SPR BLI verified) (CDA-C82E5) is expressed from human 293 cells (HEK293). It contains AA Gln 28 - Pro 208 (Accession Q8SPW4-1). 1 * 25 µG
Lieferant: ACROBIOSYSTEMS INC