Drucken
Human CD38, Llama IgG2b Fc Tag, low endotoxin (CD8-H5252) is expressed from human 293 cells (HEK293). It contains AA Val 43 - Ile 30 0 (Accession P28907-1). 1 * 50 µG
Lieferant: ACROBIOSYSTEMS INC