Human CD20 Full Length, His Tag (HEK293) (SPR verified) (CD0-H52H3) is expressed from human 293 cells (HEK293). It contains AA Met 1 - Pro 297 (Accession P11836-1). 1 * 20 µG
Lieferant: ACROBIOSYSTEMS INC