Recombinat Protein,Human Adiponectin ADIPOQ Protein, Fc Tag,Human ADIPOQ, Fc Tag (ADQ-H5250) is expressed from human 293 cells (HEK 293). It contains AA Glu 19 - Asn 244 (Accession NP001171271).This protein carries a human IgG1 Fc tag at the C-termi 1 * 100 µG
Lieferant: ACROBIOSYSTEMS INC