Anti-ELANE Mouse Monoclonal Antibody

Lieferant: Abnova
ABNOMAB2205EA 399 EUR
ABNOMAB2205
Anti-ELANE Mouse Monoclonal Antibody
Antikörper
Mouse monoclonal antibody raised against native ELANE.

Type: Primary
Antigen: ELANE
Clonality: Monoclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG2b
Reactivity: Human
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR