Anti-CA10 Mouse Monoclonal Antibody [clone: 1A10]

Lieferant: Abnova
ABNOH00056934-M01EA 394 EUR
ABNOH00056934-M01
Anti-CA10 Mouse Monoclonal Antibody [clone: 1A10]
Antikörper
Type: Primary
Antigen: CA10
Clonality: Monoclonal
Clone: 1A10
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype:
Reactivity: Human
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR