Anti-DEDD Mouse Monoclonal Antibody [clone: 1C7]

Lieferant: Abnova
ABNOH00009191-M01EA 394 EUR
ABNOH00009191-M01
Anti-DEDD Mouse Monoclonal Antibody [clone: 1C7]
Antikörper
Type: Primary
Antigen: DEDD
Clonality: Monoclonal
Clone: 1C7
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype:
Reactivity: Human
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR