Amino acid standard, AAS18, analytical standard, Supelco®
Lieferant: Merck
AAS18-10X1MLEA 185 EUR
AAS18-10X1ML AAS18-5X5ML AAS18-5ML AAS18-10ML
Amino acid standard, AAS18, analytical standard, Supelco®
Standards Lebensmittel-Standards
Amino acids (17) in this standard are 2,5 μmoles per ml in 0,1 N HCl, except L-cystine at 1,25 μmoles per ml.
The following components are included in the amino acid standard:
L-Alanine; Ammonium chloride; L-Arginine; L-Aspartic acid; L-Cystine; L-Glutamic acid; Glycine; L-Histidine; L-Isoleucine; L-Leucine; L-Lysine; L-Methionine; L-Phenylalanine; L-Proline; L-Serine; L-Threonine; L-Tyrosine; L-Valine