Amino acid standards, physiological, basics, analytical standard, Supelco®
Lieferant: Merck
A6282-5MLEA 119 EUR
A6282-5ML A6282-10ML A6282-1ML
Amino acid standards, physiological, basics, analytical standard, Supelco®
Standards Lebensmittel-Standards
This solution contains physiological, basic amino acids and related compounds for calibration of amino acid analysers.
Components of the standard:
γ-Amino-n-butyric Acid; Ammonium Chloride; L-Anserine; L-Arginine; L-Carnosine; Creatinine; Ethanolamine; L-Histidine; L-Homocystine; d-DL-Hydroxylysine; L-Lysine; 1-Methyl-L-histidine; 3-Methyl-L-histidine; L-Ornithine; L-Tryptophan