(1,3-Dioxolan-2-ylmethyl)triphenylphosphoniumbromid ≥98%

Lieferant: Thermo Scientific
Achtung

A18882.30EA 744.64 EUR
A18882.30 A18882.18 A18882.09
(1,3-Dioxolan-2-ylmethyl)triphenylphosphoniumbromid ≥98%
(1,3-Dioxolan-2-ylmethyl)triphenylphosphoniumbromid
Formel: C₂₂H₂₂BrO₂P
Molecular Weight: 429,30
Schmelzpunkt: 186…189 °C
MDL: MFCD00011966
CAS-Nummer: 52509-14-5
EINECS: 257-977-3

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR