L(-)-Fucose 98%

Lieferant: Thermo Scientific

A16789.06EA 167 EUR
A16789.06 A16789.03
L(-)-Fucose 98%
L(-)-Fucose
Formel: C₆H₁₂O₅
Molecular Weight: 164,16 g/mol
Schmelzpunkt: 139…142 °C
Lagertemperatur: Raumtemperatur
MDL: MFCD00069812
CAS-Nummer: 2438-80-4
EINECS: 219-452-7

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR