Inosin ≥98%

Lieferant: Thermo Scientific

A14459.14EA 71.4 EUR
A14459.14 A14459.22 A14459.06
Inosin ≥98%
Inosin

Applications: Suppresses the increase of glucose and insulin used in blood.

Formel: C₁₀H₁₂N₄O₅
Molecular Weight: 268,23 g/mol
Schmelzpunkt: 212…215 °C (Zersetzung)
Lagertemperatur: Raumtemperatur
MDL: MFCD00066770
CAS-Nummer: 58-63-9
EINECS: 200-390-4

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR