Micro particles based on melamine resin, analytical standard, Supelco®
Lieferant: Merck
91329-10ML-FEA 372 EUR
91329-10ML-F 95523-5ML-F 96062-10ML-F
Micro particles based on melamine resin, analytical standard, Supelco®
Standards Partikelgrößenanalyse-Standards
For every lot, exact values of particle size and standard deviation are determined with an accuracy of 0,01 μm using a Coulter Multisizer.