VWR®, Redox (ORP) Standards
Lieferant: Avantor
VWR®
91227.270EA 75.6 EUR
91227.270 91229.270 91231.270 91224.270 91235.270 91233.270 91226.270 91228.270 91225.270 91230.270 91223.270 91232.270 91234.270
VWR®, Redox (ORP) Standards
Standards Redox-/ORP-Standards
Avantor offers a wide range of REDOX (ORP) standards, which are produced from high quality reagents.These reference materials can be used as control standards to check the functionality of both the elextrodes and the measuring instruments. The values are temperature dependent, so it is important to control the environment of the measurement. A table of temperatue and corresponding Redox value is available on both the label and the batch specific certificate.
  • Accuracy ±5 mV
  • Quoted values tested at 25 °C
  • Temperature vs Redox value as a table on the bottle label
  • Stability excellent throughout shelf life
ORP standards where all values quoted are potentials of platinum electrode v Ag/AgCl reference electrode (3M KCl)