Wasser Ph. Eur., USP, JP, gereinigt

Lieferant: VWR Chemicals

Synonyme: WasserstoffoxidHydrogenoxid

90200.0999EA 0 EUR
90200.0999 90200.5000 90200.9010 90200.9200 90200.9025
Wasser Ph. Eur., USP, JP, gereinigt
Wasser

  • Produced by reverse osmosis and capacitive deionization (CDI)


Bitte beachten Not for use as Active Pharmaceutical Ingredient (API)

Formel: H₂O
Molecular Weight: 18,02 g/mol
Siedepunkt: 100 °C (1013 hPa)
Schmelzpunkt: 0 °C
Dichte: 1,00 g/cm³ (20 °C)
Lagertemperatur: Raumtemperatur
MDL: MFCD00011332
CAS-Nummer: 7732-18-5
EINECS: 231-791-2
Merck Index: 12,10175

Bestellen

Spezifikation Testergebnisse

V (Vanadium) Max. 0.1 ppm
Conforms to Ph.Eur./USP/JP Passes test
Conforms to Ph.Eur. Monograph 0008 Passes test
Ph.Eur. specifications:
Total organic carbon (TOC) [Ph.Eur.] Max. 0.5 mg/l
Conductivity [Ph.Eur.] Max. 4.3 μS/cm
Elemental impurities (by conductivity) Passes test
Conductivity (20°C) (on line) [Ph.Eur.] Max. 1.1 μS/cm
NO3 (Nitrate) [Ph.Eur.] Max. 0.2 ppm
Acidity or alkalinity Passes test Ph.Eur.
Oxidisable substances Passes test Ph.Eur.
Cl (Chloride) Passes test Ph.Eur.
SO4 (Sulphate) Passes test Ph.Eur.
NH4 (Ammonium) [Ph.Eur.] Max. 0.2 ppm
Calcium and magnesium Passes test Ph.Eur.
Residue on evaporation [Ph.Eur.] Max. 0.001 %
Microbial contamination Passes test Ph.Eur.
TAMC [Ph.Eur.] Max. 100 CFU/ml
USP specifications:
Total organic carbon [USP] Max. 0.50 mg/l
Water conductivity [USP] Max. 1.1 μS/cm
JP specifications:
Total organic carbon [JP] Max. 0.50 mg/l
Conductivity (25°C) [JP] Max. 2.1 μS/cm
KMnO4 reducing substances Passes test JP
Microbial limit Passes test JP
TAMC [JP] Max. 100 CFU/ml
Additional information
Bacterial endotoxins (Max. 0,25 IU/ml), tested on finished product Passes test

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR