Micro particles based on melamine resin, carboxylate-modified FITC-marked, analytical standard, Supelco®
Lieferant: Merck
90287-5ML-FEA 548 EUR
90287-5ML-F 88486-5ML-F
Micro particles based on melamine resin, carboxylate-modified FITC-marked, analytical standard, Supelco®
Standards Partikelgrößenanalyse-Standards
For every lot exact values of particle size and standard deviation are determined with an accuracy of 0,01 μm using a Coulter Multisizer.
Surface coverage: Reactive groups (COOH) >0,1 mmol/g polymer.