Ammoniaklösung 20 - 22% HiPerSolv CHROMANORM® für die LC-MS

Lieferant: VWR Chemicals
Gefahr

Synonyme: AmmoniakAmmoniumhydroxid

Unter Stickstoff abgepackt

88439.154EA 82 EUR
88439.154
Ammoniaklösung 20 - 22% HiPerSolv CHROMANORM® für die LC-MS
Ammoniaklösung
Formel: NH₃
Molecular Weight: 35,05 g/mol
MDL: MFCD00066650
CAS-Nummer: 1336-21-6
UN: 2672
ADR: 8,III

Bestellen

Spezifikation Testergebnisse

Assay (GC) 20 - 22%
Evaporation residue max. 0,0002%
Ca (Calcium) max. 2 ppb
K (Potassium) max. 2 ppb
Mg (Magnesium) max. 2 ppb
Na (Sodium) max. 10 ppb
Suitable for LC-MS Passes test
Manufactured and filled under nitrogen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR