Silbernitrat Reag. Ph. Eur. 1078305

Gefahr

Synonyme: Höllenstein

87927.180EA 242 EUR
87927.180
Silbernitrat Reag. Ph. Eur. 1078305
Silbernitrat

Starting materials: concentrated ammonia R / 1 M sodium hydroxide / silver nitrate R / water R


Bestellinformation: Geliefert mit Analysenzertifikat. Herstellung erfolgt nach Auftragserteilung. Bitte wenden Sie sich an Ihr lokales Vertriebszentrum.

Formel: AgNO₃
Molecular Weight: 169,87 g/mol
MDL: MFCD00003414
CAS-Nummer: 7761-88-8
UN: 3262
ADR: 8,III

For metal trace analysis in AAS or ICP

Ask for your “customized multi-element standard solution” with your exact composition using the request form

Ask for a quotation

12/31/2022

Bestellen

For metal trace analysis in AAS or ICP

Ask for your “customized multi-element standard solution” with your exact composition using the request form

Ask for a quotation

12/31/2022

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR