Organic reference standard, Methyl tetracosanoate, analytical standard, Supelco®
Lieferant: Merck
SUPL87115-1GEA 189 EUR
SUPL87115-1G SUPL87115-250MG
Organic reference standard, Methyl tetracosanoate, analytical standard, Supelco®
Methyl tetracosanoate may be used as an external standard to study the pentafluorobenzylation reaction of estradiol in a biphasic methylene chloride/sodium hydroxide system and in the absence of phase transfer agents by gas chromatography technique(GC).
  • 1 g in poly bottle
  • 250 mg in poly bottle