Phenoxyethanol ≥99,0%, GPR RECTAPUR®

Lieferant: VWR Chemicals
Achtung

Synonyme: β-HydroxyphenetolPhenylcellosolveEthylenglycolmonophenyletherω-HydroxyphenetolPhenylglycolMonophenylglycol

87105.290EA 66.4 EUR
87105.290 87105.460 87105.320
Phenoxyethanol ≥99,0%, GPR RECTAPUR®
Phenoxyethanol
Formel: C₆H₅OCH₂CH₂OH
Molecular Weight: 138,17 g/mol
MDL: MFCD00002857
CAS-Nummer: 122-99-6

Bestellen

Spezifikation Testergebnisse

Assay (GC, area%) min. 99,0%
Identification (IR) passes test
Density (20/4) 1,105 - 1,110
Peroxides (as H₂O₂) max. 50 ppm

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR