Essigsäure ≥99,8% (durch HPLC), HiPerSolv CHROMANORM® für die HPLC

Lieferant: VWR Chemicals
Gefahr

Synonyme: Ethansäure

87023.290EA 52.56 EUR
87023.290 87023.260
Essigsäure ≥99,8% (durch HPLC), HiPerSolv CHROMANORM® für die HPLC
Essigsäure
Formel: H₃CCOOH
Molecular Weight: 60,05 g/mol
MDL: MFCD00036152
CAS-Nummer: 64-19-7
UN: 2789
ADR: 8,II
https://de.vwr.com/stibo/web/std.lang.all/31/15/723115.jpg

For metal trace analysis in AAS or ICP

Ask for your “customized multi-element standard solution” with your exact composition using the request form

Ask for a quotation

12/31/2022

Bestellen

Spezifikation Testergebnisse

Assay (on anhydrous substance) Min. 99.8 %
Evaporation residue Max. 0.0001 %
Water Max. 0.1 %
Transmittance (252 nm) Min.10 %
Transmittance (260 nm) Min. 50 %
Transmittance (270 nm) Min. 80 %
Transmittance (280 nm) Min. 95 %
Transmittance (300 nm) Min. 99 %

For metal trace analysis in AAS or ICP

Ask for your “customized multi-element standard solution” with your exact composition using the request form

Ask for a quotation

12/31/2022

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR