2-Propanol ≥70% (v/v), GPR RECTAPUR®

Lieferant: VWR Chemicals

Synonyme: 2-HydroxypropanIPADimethylcarbinolIsopropanolIsopropylalkohol

86123.290EA 26.8 EUR
86123.290 86123.410 86123.460 86123.360
2-Propanol ≥70% (v/v), GPR RECTAPUR®
2-Propanol
Formel: (CH₃)₂CHOH
Molecular Weight: 60,1 g/mol
MDL: MFCD00011674
CAS-Nummer: 67-63-0
https://de.vwr.com/stibo/web/std.lang.all/19/69/731969.jpg

Bestellen

Spezifikation Testergebnisse

Assay (V/V) 68 --> 72 %
Density (20/4) 0.869 --> 0.883
n 20/D 1.137 --> 1.375
Evaporation residue Max. 20 ppm
Methanol Max. 0.1 %
Fe (Iron) Max. 1 ppm
Pb (Lead) Max. 5 ppm

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR