Chloroform ≥99,9% (GC, bezogen auf die Trockenmasse) stabilisiert, PESTINORM® Supra Trace

Lieferant: VWR Chemicals
Gefahr

Synonyme: Trichlormethan

Stabilisierung: Stabilisiert mit Amylen 0,003…0,006 % w/w
Filtriert über 0,2 µm-Filter. Verpackt unter Schutzgas.

85853.320EA 153 EUR
85853.320 85853.290
Chloroform ≥99,9% (GC, bezogen auf die Trockenmasse) stabilisiert, PESTINORM® Supra Trace
Chloroform
Formel: CHCl₃
Molecular Weight: 119,38 g/mol
Siedepunkt: 61,7 °C (1013 hPa)
Schmelzpunkt: –63 °C
Dichte: 1,4832 g/cm³ (20 °C)
Lagertemperatur: Raumtemperatur
MDL: MFCD00000826
CAS-Nummer: 67-66-3
EINECS: 200-663-8
UN: 1888
ADR: 6.1,III
REACH: 01-2119486657-20
https://de.vwr.com/stibo/web/std.lang.all/69/36/726936.jpg

For your Gaz or HPLC chromatography method

ask for your “customized multi-element standard solution” with your exact composition using the request form

Ask for a quotation

Bestellen

Spezifikation Testergebnisse

Appearance Clear colorless liquid
Color (APHA, Pt-Co) max. 10
Assay (GC, on anhydrous basis) min. 99.9 %
Acidity (as HCl) max. 0.0005 %
Residue after evaporation max. 0.0003 % w/w
Stabilizer (Amylene) 0.003-0.006 %w/w
Water (KF) max. 0.01 %w/w
GC/ECD Dioxins, Furans & PCB's max. 5 pg/ml
GC/ECD any Pesticide (as Lindane) max. 5 pg/ml
GC/NPD any Pesticide (as Parathion) max. 10 pg/ml
GC/ECD 1,2,4-TCB to daca-PCB (as Lindane) max. 5 pg/ml
GC/FID C10 to C40 (as n-Decane) max. 3 ng/ml
GC/MSD C10 to C40 (as n-Decane; 30-600amu) max. 3 ng/ml

For your Gaz or HPLC chromatography method

ask for your “customized multi-element standard solution” with your exact composition using the request form

Ask for a quotation

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR