n-Hexan ≥99%, HiPerSolv CHROMANORM® für die LC-MS

Lieferant: VWR Chemicals
Gefahr


Unter Stickstoff abgepackt

85799.320EA 219 EUR
85799.320 85799.290
n-Hexan ≥99%, HiPerSolv CHROMANORM® für die LC-MS
n-Hexan
Formel: H₃C(CH₂)₄CH₃
Molecular Weight: 86,18 g/mol
Siedepunkt: 69 °C (1013 hPa)
Schmelzpunkt: –94,3 °C
Dichte: 0,659 g/cm³ (20 °C)
Lagertemperatur: Raumtemperatur
MDL: MFCD00009520
CAS-Nummer: 110-54-3
UN: 1208
ADR: 3,
https://de.vwr.com/stibo/web/std.lang.all/70/18/737018.jpg

Bestellen

Spezifikation Testergebnisse

Assay (calculated on anhydrous) ≥ 99 %
Acidity ≤ 0.0002 meq/g
Alkalinity ≤ 0.0002 meq/g
Evaporation residue ≤ 1 ppm
Water ≤ 50 ppm
Ag (Silver) ≤ 10 ppb
Al (Aluminium) ≤ 25 ppb
Ca (Calcium) ≤ 25 ppb
Cu (Copper) ≤ 10 ppb
Fe (Iron) ≤ 10 ppb
K (Potassium) ≤ 10 ppb
Mg (Magnesium) ≤ 10 ppb
Mn (Manganese) ≤ 10 ppb
Na (Sodium) ≤ 50 ppb
Ni (Nickel) ≤ 10 ppb
Pb (Lead) ≤ 10 ppb
Zn (Zinc) ≤ 10 ppb
Transmittance (190 nm) ≥ 10 %
Transmittance (205 nm) ≥ 50 %
Transmittance (220 nm) ≥ 80 %
Transmittance (235 nm) ≥ 95 %
Transmittance (255 nm) ≥ 99 %
Packaged under nitrogen Confirmed

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR