Total Iron Test Strips, Dosatest, VWR®
Lieferant: VWR Chemicals
Dosatest
85709.601EA 8 EUR
85709.601 85713.601
Total Iron Test Strips, Dosatest, VWR®
Umwelt-Prüfkits
Semi-quantitative test strips for the determination of iron.
  • Ideal for rapid and easy analysis of iron directly on-site
  • No maintenance or calibration is required
Colour change 0 - 1000: White to dark red.
Colour change 0 - 100: White to blue violet
Lieferumfang: Delivered as 100 test strips in a tube with instruction leaflet.
Bitte beachten For the lower concentration test strip, the coloration produced by some heavy metals in higher concentrations can be eliminated by adding cadmium powder.