Ammoniaklösung 25 - 27%, NORMATOM® für die Spurenanalyse

Lieferant: VWR Chemicals

Synonyme: AmmoniakAmmoniumhydroxid

85693.290EA 170 EUR
85693.290 85693.270
Ammoniaklösung 25 - 27%, NORMATOM® für die Spurenanalyse
Ammoniaklösung
Formel: NH₃
Molecular Weight: 35,05 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD00066650
CAS-Nummer: 1336-21-6
UN: 2672
ADR: 8,III

For metal trace analysis in AAS or ICP

Ask for your “customized multi-element standard solution” with your exact composition using the request form

Ask for a quotation

12/31/2022

Bestellen

Spezifikation Testergebnisse

Assay (NH3) 25 - 27% w/w
Colouration max. 10 APHA
Cl (Chloride) max. 0.5 ppm
PO4 (Phosphate) max. 0.01 ppm
SO4 (Sulphate) max. 1 ppm
Ag (Silver) max. 0.5 ppb
Al (Aluminum) max. 1 ppb
As (Arsenic) max. 1 ppb
Au (Gold) max. 0.5 ppb
Ba (Barium) max. 0.1 ppb
Be (Beryllium) max. 0.1 ppb
Bi (Bismuth) max. 0.1 ppb
Ca (Calcium) max. 1 ppb
Cd (Cadmium) max. 0.5 ppb
Ce (Cerium) max. 0.1 ppb
Co (Cobalt) max. 0.5 ppb
Cr (Chromium) max. 0.5 ppb
Cs (Cesium) max. 0.1 ppb
Cu (Copper) max. 0.5 ppb
Dy (Dysprosium) max. 0.1 ppb
Er (Erbium) max. 0.1 ppb
Eu (Europium) max. 0.1 ppb
Fe (Iron) max. 1 ppb
Ga (Gallium) max. 0.1 ppb
Gd (Gadolinium) max. 0.1 ppb
Ge (Germanium) max. 0.1 ppb
Hf (Hafnium) Information Only
Hg (Mercury) max. 0.2 ppb
Ho (Holmium) max. 0.1 ppb
In (Indium) max. 0.1 ppb
K (Potassium) max. 1 ppb
La (Lanthanum) max. 0.1 ppb
Li (Lithium) max. 0.1 ppb
Lu (Lutetium) max. 0.1 ppb
Mg (Magnesium) max. 1 ppb
Mn (Manganese) max. 0.5 ppb
Mo (Molybdenum) max. 0.5 ppb
Na (Sodium) max. 1 ppb
Nb (Niobium) max. 0.1 ppb
Nd (Neodymium) max. 0.1 ppb
Ni (Nickel) max. 0.5 ppb
Pb (Lead) max. 0.1 ppb
Pd (Palladium) Information Only
Pr (Praseodymium) max. 0.1 ppb
Pt (Platinum) Information Only
Rb (Rubidium) max. 0.1 ppb
Re (Rhenium) Information Only
Rh (Rhodium) max. 0.5 ppb
Ru (Ruthenium) Information Only
Sb (Antimony) max. 0.5 ppb
Sc (Scandium) max. 0.1 ppb
Se (Selenium) max. 1 ppb
Sm (Samarium) max. 0.1 ppb
Sn (Tin) max. 0.5 ppb
Sr (Strontium) max. 0.1 ppb
Tb (Terbium) max. 0.1 ppb
Te (Tellurium) max. 0.1 ppb
Th (Thorium) max. 0.1 ppb
Ti (Titanium) max. 0.5 ppb
Tl (Thallium) max. 0.1 ppb
Tm (Thulium) max. 0.1 ppb
U (Uranium) max. 0.1 ppb
V (Vanadium) max. 0.5 ppb
W (Tungsten) max. 0.1 ppb
Y (Yttrium) max. 0.1 ppb
Yb (Ytterbium) max. 0.1 ppb
Zn (Zinc) max. 0.5 ppb
Zr (Zirconium) max. 0.1 ppb

For metal trace analysis in AAS or ICP

Ask for your “customized multi-element standard solution” with your exact composition using the request form

Ask for a quotation

12/31/2022

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR