Natriumhydroxid-Lösung 10%, AnalaR NORMAPUR®

Lieferant: VWR Chemicals

85660.290EA 38.3 EUR
85660.290 85660.410
Natriumhydroxid-Lösung 10%, AnalaR NORMAPUR®
Sodium hydroxide
Formel: NaOH
Molecular Weight: 40 g/mol
MDL: MFCD00003548
CAS-Nummer: 1310-73-2

Bestellen

Spezifikation Testergebnisse

Assay 10.0 - 10.5%
Heavy metals (as Pb) max. 5 ppm
Total nitrogen (N) max. 1 ppm
Carbonate (as Na₂CO₃) max. 1%
Chloride (Cl) max. 5 ppm
Phosphate (PO₄) max. 5 ppm
Silicate (as SiO₂) max. 10 ppm
Sulphate (SO₄) max. 5 ppm
Aluminium (Al) max. 5 ppm
Iron (Fe) max. 5 ppm

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR