pH buffers, for Karl Fischer titrations, TITRINORM®
Lieferant: VWR, part of Avantor
85498.290EA 134 EUR
85498.290DP 85498.290 85497.260
pH buffers, for Karl Fischer titrations, TITRINORM®
Karl Fischer Titriermittel und Reagenzien
The volumetric and coulometric reagents are buffered systems, stabilised around the ideal value between 5 to 7 ensuring a rapid and stoichiometric course of reaction. Buffering systems which neutralise and stabilise the pH during the reaction are suitable for this purpose.