VWR® TITRINORM®, pH Buffers, for Karl Fischer Titrations
Lieferant: Avantor
85498.290EA 141 EUR
85498.290DP 85498.290 85497.260
VWR® TITRINORM®, pH Buffers, for Karl Fischer Titrations
Karl Fischer Titriermittel und Reagenzien
The volumetric and coulometric reagents are buffered systems, stabilised around the ideal value between 5 to 7 ensuring a rapid and stoichiometric course of reaction. Buffering systems which neutralise and stabilise the pH during the reaction are suitable for this purpose.